- 4-thiazolidinone--a biologically active scaffold.
4-thiazolidinone--a biologically active scaffold.
European journal of medicinal chemistry (2007-09-18)
Amit Verma, Shailendra K Saraf
PMID17870209
ABSTRACT
The broad and potent activity of 4-thiazolidinones has established it as one of the biologically important scaffolds. This article is an effort to highlight the importance of the 4-thiazolidinones in the present context and promise they hold for the future.