MilliporeSigma
  • Sublingual immunotherapy alters salivary IgA and systemic immune mediators in timothy allergic children.
Sublingual immunotherapy alters salivary IgA and systemic immune mediators in timothy allergic children.
Pediatric allergy and immunology : official publication of the European Society of Pediatric Allergy and Immunology (2019-02-26)
Johanna Huoman, Georgia Papapavlou, Anna Pap, Johan Alm, Lennart J Nilsson, Maria C Jenmalm
ABSTRACT
Immunomodulatory effects of sublingual immunotherapy on systemic and mucosal mediators in allergic children are largely unexplored. The aim of this study was to investigate allergy-related cytokine and chemokine levels, as well as IgA-responses upon a 3-year treatment with timothy grass pollen sublingual immunotherapy in children with allergic rhinoconjunctivitis. From children included in the GRAZAX® Asthma Prevention study, blood and saliva samples were analyzed at inclusion, after 3 years of treatment, and 2 years after treatment ending. By means of Luminex and ELISA methodologies, allergy-related cytokines and chemokines were measured in plasma samples and allergen-stimulated peripheral blood mononuclear cell supernatants. Furthermore, studies of total, secretory, and Phl p 1-specific salivary IgA antibodies were performed using the same methods. GRAZAX® -treated children exhibited significantly higher levels of Phl p 1-specific salivary IgA and serum IgG4 , along with significantly lower skin prick test positivity, after 3 years of treatment and 2 years after treatment cessation. Additionally, plasma levels of the Th1-associated chemokines CXCL10 and CXCL11 were significantly higher in treated than untreated children at these time points. Timothy-induced ratios of IL-5/IL-13 over IFN-γ were significantly decreased after 3 years with active treatment, as were symptoms of allergic rhinitis in terms of both severity and visual analogue scale scores. However, no consistent correlations were found between the clinical outcomes and immunologic parameters. Phleum pratense sublingual immunotherapy in grass pollen allergic children modulates the immune response in the oral mucosa as well as systemically-by increasing Th1-responses, decreasing Th2-responses, and inducing immunoregulatory responses-all signs of tolerance induction.
Sigma-Aldrich
IgA from human colostrum, reagent grade, ≥95%, essentially salt-free, lyophilized powder
Sigma-Aldrich
Anti-Human IgA (α-chain specific)−Peroxidase antibody produced in goat, affinity isolated antibody