MilliporeSigma
  • Human corneal fibroblast migration and extracellular matrix synthesis during stromal repair: Role played by platelet-derived growth factor-BB, basic fibroblast growth factor, and transforming growth factor-β1.
Human corneal fibroblast migration and extracellular matrix synthesis during stromal repair: Role played by platelet-derived growth factor-BB, basic fibroblast growth factor, and transforming growth factor-β1.
Journal of tissue engineering and regenerative medicine (2016-11-20)
Patricia Gallego-Muñoz, Lucía Ibares-Frías, José A Garrote, María Cruz Valsero-Blanco, Roberto Cantalapiedra-Rodríguez, Jesús Merayo-Lloves, M Carmen Martínez-García
ABSTRACT
The development of treatments that modulate corneal wound healing to avoid fibrosis during tissue repair is important for the restoration of corneal transparency after an injury. To date, few studies have studied the influence of growth factors (GFs) on human corneal fibroblast (HCF) expression of extracellular matrix (ECM) proteins such as collagen types I and III, proteoglycans such as perlecan, or proteins implicated in cellular migration such as α5β1-integrin and syndecan-4. Using in vitro HCFs, a mechanical wound model was developed to study the influence of the GFs basic fibroblast GF (bFGF), platelet-derived GF (PDGF-BB) and transforming GF-β1 (TGFβ1) on ECM protein production and cellular migration. Our results show that mechanical wounding provokes the autocrine release of bFGF and TGFβ1 at different time points during the wound closure. The HCF response to PDGF-BB was a rapid closure due to fast cellular migration associated with a high focal adhesion replacement and a high expression of collagen and proteoglycans, producing nonfibrotic healing. bFGF stimulated nonfibrotic ECM production and limited the migration process. Finally, TGFβ1 induced expression of the fibrotic markers collagen type III and α5β1 integrin, and it inhibited cellular migration due to the formation of focal adhesions with a low turnover rate. The novel in vitro HCF mechanical wound model can be used to understand the role played by GFs in human corneal repair. The model can also be used to test the effects of different treatments aimed at improving the healing process. Copyright © 2016 John Wiley & Sons, Ltd.
Sigma-Aldrich
Anti-Collagen Type I Rabbit pAb, liquid, Calbiochem®