MilliporeSigma
  • Repeated exposure to heat stress induces mitochondrial adaptation in human skeletal muscle.
Repeated exposure to heat stress induces mitochondrial adaptation in human skeletal muscle.
Journal of applied physiology (Bethesda, Md. : 1985) (2018-07-20)
Paul S Hafen, Coray N Preece, Jacob R Sorensen, Chad R Hancock, Robert D Hyldahl
ABSTRACT
The heat stress response is associated with several beneficial adaptations that promote cell health and survival. Specifically, in vitro and animal investigations suggest that repeated exposures to a mild heat stress (~40°C) elicit positive mitochondrial adaptations in skeletal muscle comparable to those observed with exercise. To assess whether such adaptations translate to human skeletal muscle, we produced local, deep tissue heating of the vastus lateralis via pulsed shortwave diathermy in 20 men and women ( n = 10 men; n = 10 women). Diathermy increased muscle temperature by 3.9°C within 30 min of application. Immediately following a single 2-h heating session, we observed increased phosphorylation of AMP-activated protein kinase and ERK1/2 but not of p38 MAPK or JNK. Following repeated heat exposures (2 h daily for 6 consecutive days), we observed a significant cellular heat stress response, as heat shock protein 70 and 90 increased 45% and 38%, respectively. In addition, peroxisome proliferator-activated receptor gamma, coactivator-1 alpha and mitochondrial electron transport protein complexes I and V expression were increased after heating. These increases were accompanied by augmentation of maximal coupled and uncoupled respiratory capacity, measured via high-resolution respirometry. Our data provide the first evidence that mitochondrial adaptation can be elicited in human skeletal muscle in response to repeated exposures to mild heat stress. NEW & NOTEWORTHY Heat stress has been shown to elicit mitochondrial adaptations in cell culture and animal research. We used pulsed shortwave diathermy to produce deep tissue heating and explore whether beneficial mitochondrial adaptations would translate to human skeletal muscle in vivo. We report, for the first time, positive mitochondrial adaptations in human skeletal muscle following recurrent heat stress. The results of this study have clinical implications for many conditions characterized by diminished skeletal muscle mitochondrial function.
Product Description
Millipore
MILLIPLEX MAP Multi-Pathway Magnetic Bead 9-Plex - Cell Signaling Multiplex Assay