MilliporeSigma
  • First epidemiological study on exposure to Neospora caninum in different canine populations in the Algiers District (Algeria).
First epidemiological study on exposure to Neospora caninum in different canine populations in the Algiers District (Algeria).
Parasitology international (2009-09-03)
F Ghalmi, B China, R Kaidi, B Losson
ABSTRACT
Neospora caninum is an important cause of abortion in cattle worldwide. Dogs act as final hosts shedding oocysts in the environment. They can also harbour the extraintestinal stage of the parasite and this may be associated with a fairly rare neuromuscular condition. The sera of 781 dogs from the Algiers District were screened by IFAT for the presence of anti-N. caninum antibodies. These dogs were distributed into four populations: local stray dogs, police dogs, dogs from breeding kennels and farm dogs. The overall seroprevalence was 21.90%. Significant differences were observed between the different populations, the highest prevalence being observed in farm (44.44%) and stray dogs (22.55%). Additionally, the highest titres were observed in farm dogs. Among studied epidemiological parameters, breed, dog origin, season and vaccination status were significantly associated with IFAT results. Additionally, a recently described real time PCR was used on the blood of 100 pound dogs and the results were compared with the serological data. A higher proportion of dogs was found to be positive by PCR when compared to the IFAT results. There was only a fairly low agreement between PCR and IFAT results which suggests that these techniques measured different aspects of the host-parasite relationship. This study indicates that the level of exposure of the canine population of Algiers area to N. caninum is very high. This would indicate a potentially high risk for N. caninum induced abortion in cattle in this region and in Algeria.
Sigma-Aldrich
Anti-Dog IgG (whole molecule)−Peroxidase antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution