MilliporeSigma
  • Variability of NOR patterns in European water frogs of different genome composition and ploidy level.
Variability of NOR patterns in European water frogs of different genome composition and ploidy level.
Comparative cytogenetics (2017-09-19)
Anna Zaleśna, Maria Florek, Mariusz Rybacki, Maria Ogielska
ABSTRACT
We studied water frogs from a complex composed of two species: Pelophylax lessonae (Camerano, 1882) (genome LL, 2n = 26) and P. ridibundus (Pallas, 1771) (RR, 2 = 26), and their natural hybrid P. esculentus (Fitzinger, 1843) of various ploidy and genome composition (RL, 2n = 26, and RRL or RLL, 3n = 39). Tetraploids RRLL were found (4n = 52) in juveniles. We applied cytogenetic techniques: AgNO3, chromomycin A3, PI and fluorescent in situ hybridization with a 28S rDNA probe. Results obtained by silver staining corresponded well with those stained with CMA3, PI and FISH. As a rule, NORs are situated on chromosomes 10. The number of Ag-NORs visible on metaphase plates was the same as the number of Ag-nucleoli present in interphase nuclei of the same individual. In all analyzed metaphases, NORs exhibited variations in size after AgNO3 and CMA3 stainings. Sixty-six individuals (out of 407 analyzed) were polymorphic for the localization and number of NORs. Fifty-one diploids had NORs only on one chromosome of pair 10. Three triploids (LLR and RRL) displayed two NORs, and two other triploid RRL individuals displayed one, instead of expected three NORs. In ten individuals extra NORs were detected on chromosomes other than 10 (chromosomes 2 and 9).
Product Description
Roche
Nick Translation Kit, sufficient for 50 labeling reactions, kit of 1 (7 components), suitable for FISH