MilliporeSigma
  • A homogeneous bioluminescent immunoassay to probe cellular signaling pathway regulation.
A homogeneous bioluminescent immunoassay to probe cellular signaling pathway regulation.
Communications biology (2020-01-08)
Byounghoon Brian Hwang, Laurie Engel, Said A Goueli, Hicham Zegzouti
ABSTRACT
Monitoring cellular signaling events can help better understand cell behavior in health and disease. Traditional immunoassays to study proteins involved in signaling can be tedious, require multiple steps, and are not easily adaptable to high throughput screening (HTS). Here, we describe a new immunoassay approach based on bioluminescent enzyme complementation. This immunoassay takes less than two hours to complete in a homogeneous "Add and Read" format and was successfully used to monitor multiple signaling pathways' activation through specific nodes of phosphorylation (e.g pIκBα, pAKT, and pSTAT3). We also tested deactivation of these pathways with small and large molecule inhibitors and obtained the expected pharmacology. This approach does not require cell engineering. Therefore, the phosphorylation of an endogenous substrate is detected in any cell type. Our results demonstrate that this technology can be broadly adapted to streamline the analysis of signaling pathways of interest or the identification of pathway specific inhibitors.
Sigma-Aldrich
Interleukin-1β Protein, Recombinant human, The recombinant human Interleukin-1beta protein (or IL-1beta protein) is a potent immuno-modulator which mediates a wide range of immune & inflammatory responses including the activation of B & T-cells.