MilliporeSigma
  • Ligation of 2', 3'-cyclic phosphate RNAs for the identification of microRNA binding sites.
Ligation of 2', 3'-cyclic phosphate RNAs for the identification of microRNA binding sites.
FEBS letters (2020-10-29)
Christian Berk, Yuluan Wang, Artur Laski, Stylianos Tsagkris, Jonathan Hall
ABSTRACT
Identifying the targetome of a microRNA is key for understanding its functions. Cross-linking and immunoprecipitation (CLIP) methods capture native miRNA-mRNA interactions in cells. Some of these methods yield small amounts of chimeric miRNA-mRNA sequences via ligation of 5'-phosphorylated RNAs produced during the protocol. Here, we introduce chemically synthesized microRNAs (miRNAs) bearing 2'-, 3'-cyclic phosphate groups, as part of a new CLIP method that does not require 5'-phosphorylation for ligation. We show in a system that models miRNAs bound to their targets, that addition of recombinant bacterial ligase RtcB increases ligation efficiency, and that the transformation proceeds via a 3'-phosphate intermediate. By optimizing the chemistry underlying ligation, we provide the basis for an improved method to identify miRNA targetomes.
Sigma-Aldrich
Water-18O, (for PET), 97 atom % 18O