MilliporeSigma
  • Design and pharmacophore modeling of biaryl methyl eugenol analogs as breast cancer invasion inhibitors.
Design and pharmacophore modeling of biaryl methyl eugenol analogs as breast cancer invasion inhibitors.
Bioorganic & medicinal chemistry (2009-12-26)
Fatma M Abdel Bar, Mohammad A Khanfar, Ahmed Y Elnagar, Farid A Badria, Ahmed M Zaghloul, Kadria F Ahmad, Paul W Sylvester, Khalid A El Sayed
ABSTRACT
Cell invasion and migration are required for the parent solid tumor cells to metastasize to distant organs. Microtubules form a polarized network, enabling organelle and protein movement throughout the cell. Cytoskeletal elements coordinately regulate cell's motility, adhesion, migration, exocytosis, endocytosis, and division. Thus, microtubule disruption can be a useful target to control cancer cell invasion and metastasis. The phenolic ether methyl eugenol (1), the major component of the essential oil of the leaves of Melaleuca ericifolia Sm. (Myrtaceae), was used as a starting scaffold to design eleven new and three known anti-tubulin agents 2-15 using carbon-carbon coupling reactions. A computer-assisted approach was used to design these new biaryl derivatives using colchicine-binding site of tubulin as the molecular target and colchicine as an active ligand. Several derivatives showed potent inhibitory activity against MDA-MB-231 cell migration at the 1-4microM dose range. The Z isomers, 4 and 15 were more active as invasion inhibitors compared to their structurally related E isomers, 2 and 14. The cytotoxic activities of compounds 2-15 against two breast cancer cell lines MDA-MB-231 and MCF-7 were evaluated. Anti-invasive activity of the semisynthetic derivatives is not due to a direct cytotoxic effect on MDA-MB-231. Analogs 2-15 may promote their anti-invasive activity through the induction of changes in cell morphology. A pharmacophore model was generated involving seven essential features for activity, which was consistent with a previously generated colchicine site inhibitors model.
Sigma-Aldrich
Colchicine, BioReagent, suitable for plant cell culture, ≥95% (HPLC)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Colchicine, ≥95% (HPLC), powder