MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Emissive spin-0 triplet-pairs are a direct product of triplet-triplet annihilation in pentacene single crystals and anthradithiophene films.
Emissive spin-0 triplet-pairs are a direct product of triplet-triplet annihilation in pentacene single crystals and anthradithiophene films.
Nature chemistry (2020-12-09)
David G Bossanyi, Maik Matthiesen, Shuangqing Wang, Joel A Smith, Rachel C Kilbride, James D Shipp, Dimitri Chekulaev, Emma Holland, John E Anthony, Jana Zaumseil, Andrew J Musser, Jenny Clark
ABSTRACT
Singlet fission and triplet-triplet annihilation represent two highly promising ways of increasing the efficiency of photovoltaic devices. Both processes are believed to be mediated by a biexcitonic triplet-pair state, 1(TT). Recently however, there has been debate over the role of 1(TT) in triplet-triplet annihilation. Here we use intensity-dependent, low-temperature photoluminescence measurements, combined with kinetic modelling, to show that distinct 1(TT) emission arises directly from triplet-triplet annihilation in high-quality pentacene single crystals and anthradithiophene (diF-TES-ADT) thin films. This work demonstrates that a real, emissive triplet-pair state acts as an intermediate in both singlet fission and triplet-triplet annihilation and that this is true for both endo- and exothermic singlet fission materials.
Sigma-Aldrich
Pentacene, triple-sublimed grade, ≥99.995% trace metals basis