MilliporeSigma
  • Amorphous Alloy Architectures in Pore Walls: Mesoporous Amorphous NiCoB Alloy Spheres with Controlled Compositions via a Chemical Reduction.
Amorphous Alloy Architectures in Pore Walls: Mesoporous Amorphous NiCoB Alloy Spheres with Controlled Compositions via a Chemical Reduction.
ACS nano (2020-12-15)
Yunqing Kang, Bo Jiang, Juanjuan Yang, Zhe Wan, Jongbeom Na, Qian Li, Hexing Li, Joel Henzie, Yoshio Sakka, Yusuke Yamauchi, Toru Asahi
ABSTRACT
Amorphous bimetallic borides are an emerging class of catalytic nanomaterial that has demonstrated excellent catalytic performance due to its glass-like structure, abundant unsaturated active sites, and synergistic electronic effects. However, the creation of mesoporous Earth-abundant bimetallic metal borides with tunable metal proportion remains a challenge. Herein, we develop a sophisticated and controllable dual-reducing agent strategy to synthesize the mesoporous nickel-cobalt boron (NiCoB) amorphous alloy spheres (AASs) with adjustable compositions by using a soft template-directed assembly approach. The selective use of tetrabutylphosphonium bromide (Bu4PBr) is beneficial to generate well-defined mesopores because it both moderates the reduction rate by decreasing the reducibility of M2+ species and prevents the generation of soap bubbles. Our meso-Ni10.0Co74.5B15.5 AASs generate the highest catalytic performance for the hydrolytic dehydrogenation of ammonia borane (AB). Its high performance is attributed to the combination of optimal synergistic effects between Ni, Co, and B as well as the high surface area and the good mass transport efficiency due to the open mesopores. This work describes a systematic approach for the design and synthesis of mesoporous bimetallic borides as efficient catalysts.
Sigma-Aldrich
Tetrabutylphosphonium bromide, 98%