MilliporeSigma
  • Mechanisms of Resistance to Ceftolozane/Tazobactam in Pseudomonas aeruginosa: Results of the GERPA Multicenter Study.
Mechanisms of Resistance to Ceftolozane/Tazobactam in Pseudomonas aeruginosa: Results of the GERPA Multicenter Study.
Antimicrobial agents and chemotherapy (2020-11-18)
Damien Fournier, Romain Carrière, Maxime Bour, Emilie Grisot, Pauline Triponney, Cédric Muller, Jérôme Lemoine, Katy Jeannot, Patrick Plésiat
ABSTRACT
Resistance mechanisms of Pseudomonas aeruginosa to ceftolozane/tazobactam (C/T) were assessed on a collection of 420 nonredundant strains nonsusceptible to ceftazidime (MIC > 8 μg/ml) and/or imipenem (>4 μg/ml), collected by 36 French hospital laboratories over a one-month period (the GERPA study). Rates of C/T resistance (MIC > 4/4 μg/ml) were equal to 10% in this population (42/420 strains), and 23.2% (26/112) among the isolates resistant to both ceftazidime and imipenem. A first group of 21 strains (50%) was found to harbor various extended-spectrum β-lactamases (1 OXA-14; 2 OXA-19; 1 OXA-35; 1 GES-9; and 3 PER-1), carbapenemases (2 GES-5; 1 IMP-8; and 8 VIM-2), or both (1 VIM-2/OXA-35 and 1 VIM-4/SHV-2a). All the strains of this group belonged to widely distributed epidemic clones (ST111, ST175, CC235, ST244, ST348, and ST654), and were highly resistant to almost all the antibiotics tested except colistin. A second group was composed of 16 (38%) isolates moderately resistant to C/T (MICs from 8/4 to 16/4 μg/ml), of which 7 were related to international clones (ST111, ST253, CC274, ST352, and ST386). As demonstrated by targeted mass spectrometry, cloxacillin-based inhibition tests, and gene bla PDC deletion experiments, this resistance phenotype was correlated with an extremely high production of cephalosporinase PDC. In part accounting for this strong PDC upregulation, genomic analyses revealed the presence of mutations in the regulator AmpR (D135N/G in 6 strains) and enzymes of the peptidoglycan recycling pathway, such as AmpD, PBP4, and Mpl (9 strains). Finally, all of the 5 (12%) remaining C/T-resistant strains (group 3) appeared to encode PDC variants with mutations known to improve the hydrolytic activity of the β-lactamase toward ceftazidime and C/T (F147L, ΔL223-Y226, E247K, and N373I). Collectively, our results highlight the importance of both intrinsic and transferable mechanisms in C/T-resistant P. aeruginosa Which mutational events lead some clinical strains to massively produce the natural cephalosporinase PDC remains incompletely understood.
Product Description
Supelco
Imipenem, Pharmaceutical Secondary Standard; Certified Reference Material
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
In Stock
Details...
$53.50
100 g
In Stock
Details...
$123.00
Sigma-Aldrich
6-Mercaptohexanoic acid, 90%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
In Stock
Details...
$53.50
100 g
In Stock
Details...
$123.00