- Clinical outcomes of 28 cats 12-24 months after urethrostomy.
Clinical outcomes of 28 cats 12-24 months after urethrostomy.
The aim of this study was to evaluate and compare the long-term clinical outcomes and quality of life of cats having undergone perineal urethrostomy (PU) or prepubic urethrostomy (PPU). This clinical study followed 28 cats (PU, n = 22; PPU, n = 6) that underwent a urethrostomy, with a minimum of 1 year postoperative follow-up. Medical records, pet owner surveys and urologic laboratory tests were used for assessment. Urologic laboratory tests included serum symmetric dimethylarginine (SDMA), serum creatinine, urinalysis, urine specific gravity (USG), urine protein:creatinine (UPC) ratio and urine culture. The main indications for urethrostomy were multiple catheterizations and PU stricture. The overall complication rates of PU and PPU were 31.8% and 83.3%, respectively. Recurrent urinary tract infection (UTI) and urine scald dermatitis were less frequent in PU than in PPU cats (UTI 22.7% vs 66.6%; dermatitis 4.5% vs 83.3%). Bacteriuria was present in 77.2% and 100% of PU and PPU cats, respectively. Owner satisfaction rates were excellent in 81.8% of PU and 33.3% of PPU cases. A proportion of cats that underwent urethrostomy showed bacteriuria, recurrent UTIs and increased levels of SDMA. PPU is important as a salvage procedure; however, it should be limited to cases in which standard techniques for PU cannot be performed, owing to the potential for recurrent complications and lower owner satisfaction.