Merck
  • Large-Scale Production of LGR5-Positive Bipotential Human Liver Stem Cells.
Large-Scale Production of LGR5-Positive Bipotential Human Liver Stem Cells.
Hepatology (Baltimore, Md.) (2019-11-13)
Kerstin Schneeberger, Natalia Sánchez-Romero, Shicheng Ye, Frank G van Steenbeek, Loes A Oosterhoff, Iris Pla Palacin, Chen Chen, Monique E van Wolferen, Gilles van Tienderen, Ruby Lieshout, Haaike Colemonts-Vroninks, Imre Schene, Ruurdtje Hoekstra, Monique M A Verstegen, Luc J W van der Laan, Louis C Penning, Sabine A Fuchs, Hans Clevers, Joery De Kock, Pedro M Baptista, Bart Spee
ABSTRACT
The gap between patients on transplant waiting lists and available donor organs is steadily increasing. Human organoids derived from leucine-rich repeat-containing G protein-coupled receptor 5 (LGR5)-positive adult stem cells represent an exciting new cell source for liver regeneration; however, culturing large numbers of organoids with current protocols is tedious and the level of hepatic differentiation is limited. Here, we established a method for the expansion of large quantities of human liver organoids in spinner flasks. Due to improved oxygenation in the spinner flasks, organoids rapidly proliferated and reached an average 40-fold cell expansion after 2 weeks, compared with 6-fold expansion in static cultures. The organoids repopulated decellularized liver discs and formed liver-like tissue. After differentiation in spinner flasks, mature hepatocyte markers were highly up-regulated compared with static organoid cultures, and cytochrome p450 activity reached levels equivalent to hepatocytes. We established a highly efficient method for culturing large numbers of LGR5-positive stem cells in the form of organoids, which paves the way for the application of organoids for tissue engineering and liver transplantation.
Product Description
Corning® disposable spinner flask, with aseptic, 1 L