MilliporeSigma
  • Tricyclic compounds containing nonenolizable cyano enones. A novel class of highly potent anti-inflammatory and cytoprotective agents.
Tricyclic compounds containing nonenolizable cyano enones. A novel class of highly potent anti-inflammatory and cytoprotective agents.
Journal of medicinal chemistry (2011-03-03)
Tadashi Honda, Hidenori Yoshizawa, Chitra Sundararajan, Emilie David, Marc J Lajoie, Frank G Favaloro, Tomasz Janosik, Xiaobo Su, Yukiko Honda, Bill D Roebuck, Gordon W Gribble
ABSTRACT
Forty-four novel tricycles containing nonenolizable cyano enones (TCEs) were designed and synthesized on the basis of a semisynthetic pentacyclic triterpenoid, bardoxolone methyl, which is currently being developed in phase II clinical trials for the treatment of severe chronic kidney disease in diabetic patients. Most of the TCEs having two different kinds of nonenolizable cyano enones in rings A and C are highly potent suppressors of induction of inducible nitric oxide synthase stimulated with interferon-γ and are highly potent inducers of the cytoprotective enzymes heme oxygenase-1 and NAD(P)H:quinone oxidoreductase-1. Among these compounds, (±)-(4bS,8aR,10aS)-10a-ethynyl-4b,8,8-trimethyl-3,7-dioxo-3,4b,7,8,8a,9,10,10a-octahydrophenanthrene-2,6-dicarbonitrile ((±)-31) is the most potent in these bioassays in our pool of drug candidates including semisynthetic triterpenoids and synthetic tricycles. These facts strongly suggest that an essential factor for potency is not a triterpenoid skeleton but the cyano enone functionality. Notably, TCE 31 reduces hepatic tumorigenesis induced with aflatoxin in rats. Further preclinical studies and detailed mechanism studies on 31 are in progress.
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Supelco
Dexamethasone, VETRANAL®, analytical standard
Sigma-Aldrich
Hydrocortisone solution, 50 μM, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Hydrocortisone-Water Soluble, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications