MilliporeSigma
  • MSN, MWCNT and ZnO nanoparticle-induced CHO-K1 cell polarisation is linked to cytoskeleton ablation.
MSN, MWCNT and ZnO nanoparticle-induced CHO-K1 cell polarisation is linked to cytoskeleton ablation.
Journal of nanobiotechnology (2021-02-14)
Karmveer Yadav, Syed Azmal Ali, Ashok Kumar Mohanty, Eshwarmoorthy Muthusamy, Kesavan Subaharan, Gautam Kaul
ABSTRACT
The cellular response to nanoparticles (NPs) for the mechanical clue and biochemical changes are unexplored. Here, we provide the comprehensive analysis of the Chinese Hamster Ovary (CHO-K1) cell line to study cell behaviour following the exposure of mesoporous silica nanoparticle (MSN), multiwall carbon nanotubes (MWCNTs), and zinc oxide (ZnO) NPs. Through the high-throughput proteomic study, we observed that the effect of NPs is alone not restricted to cell viability but also on cell polarisation. In the case of MSN, no drastic changes were observed in cellular morphology, but it upregulated chaperons that might prevent protein aggregation. However, MWCNT showed elongated cell appearance with numerous cytoplasmic vacuoles, and induce lamellipodia formation through actin polymerisation. The cytoskeleton remodelling was accompanied by the increased expression of Dlc-1, cofilin and Rac1 proteins. While ZnO NPs resulted in the rounded cell morphology along with nuclear abnormalities. The proteome analysis revealed that UBXN11 control cell roundness and DOCK3 leads to actin stress fibre formation and finally, loss of cell adhesion. It enhances the expression of catastrophic DNA damage and apoptotic proteins, which was unrecoverable even after 72 h, as confirmed by the colony formation assay. All three NPs trigger over-expression of the endocytic pathway, ubiquitination, and proteasomal complex proteins. The data indicate that ZnO and MSN entered into the cells through clathrin-mediated pathways; whereas, MWCNT invades through ER-mediated phagocytosis. Based on the incubation and concentration of NPs, our work provides evidence for the activation of Rac-Rho signalling pathway to alter cytoskeleton dynamics. Our results assist as a sensitive early molecular readout for nanosafety assessment.
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
Only 0 left in stock
$33.70
100 g
Estimated to ship on March 21, 2025
$91.00
Sigma-Aldrich
Tetraethylammonium borohydride, technical, ≥95% (T)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
Only 0 left in stock
$33.70
100 g
Estimated to ship on March 21, 2025
$91.00
Sigma-Aldrich
Zinc oxide, dispersion, nanoparticles, <100 nm particle size (TEM), ≤40 nm avg. part. size (APS), 20 wt. % in H2O
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
Only 0 left in stock
$33.70
100 g
Estimated to ship on March 21, 2025
$91.00
Sigma-Aldrich
Carbon nanotube, multi-walled, >90% carbon basis, D × L 110-170 nm × 5-9 μm
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
25 g
Only 0 left in stock
$33.70
100 g
Estimated to ship on March 21, 2025
$91.00