MilliporeSigma
  • Native bacteria isolated from roots and rhizosphere of Solanum lycopersicum L. increase tomato seedling growth under a reduced fertilization regime.
Native bacteria isolated from roots and rhizosphere of Solanum lycopersicum L. increase tomato seedling growth under a reduced fertilization regime.
Scientific reports (2020-09-26)
María Micaela Pérez-Rodriguez, Patricia Piccoli, María Soledad Anzuay, Rita Baraldi, Luisa Neri, Tania Taurian, Miguel Andrés Lobato Ureche, Diana María Segura, Ana Carmen Cohen
ABSTRACT
In semiarid regions is important to use native strains best adapted to these environments to optimize plant-PGPR interaction. We aimed to isolate and characterize PGPR from roots and rhizosphere of a tomato crop, as well as studying the effect of its inoculation on tomato seedlings growth. We selected four strains considering their effectiveness of fixing nitrogen, solubilizing phosphate, producing siderophores and indole acetic acid. They belong to the genera Enterobacter, Pseudomonas, Cellulosimicrobium, and Ochrobactrum. In addition, we also analyzed the ability to solubilize Ca3(PO4)2, FePO4 and AlPO4 and the presence of one of the genes encoding the cofactor PQQ in their genome. Enterobacter 64S1 and Pseudomonas 42P4 showed the highest phosphorus solubilizing activity and presence of pqqE gene. Furthermore, in a tomato-based bioassay in speed-bed demonstrated that a sole inoculation at seedling stage with the strains increased dry weight of roots (49-88%) and shoots (39-55%), stem height (8-13%) and diameter (5-8%) and leaf area (22-31%) and were equal or even higher than fertilization treatment. Leaf nitrogen and chlorophyll levels were also increased (50-80% and 26-33%) compared to control. These results suggest that Enterobacter 64S1 and Pseudomonas 42P4 can be used as bio-inoculant in order to realize a nutrient integrated management.
Sigma-Aldrich
Indophenol Blue, Dye content 60 %