MilliporeSigma
  • Fluorescence assay for the determination of glutathione based on a ring-fused 2-pyridone derivative in dietary supplements.
Fluorescence assay for the determination of glutathione based on a ring-fused 2-pyridone derivative in dietary supplements.
The Analyst (2021-01-23)
Wiktor Kasprzyk, Filip Koper, Agata Flis, Dominika Szreder, Elżbieta Pamuła, Dariusz Bogdał, Sławomir Wybraniec, Joanna Ortyl, Tomasz Swiergosz
ABSTRACT
Herein, a novel fluorescent method for the determination of GSH levels in aqueous solutions involving the utilization of citric acid as a derivatization reagent was developed. Therefore, the crucial parameters of the derivatization process were established from what has resulted in the development of a sensitive, reproducible, and accurate GSH assay. The method was validated, and its applicability in the characterization of the GSH concentration in dietary supplements concerning the selectivity in the determination of GSH over GSSG was both confirmed. The chemical structure of the new fluorophore 3-[(carboxymethyl)carbamoyl]-5-oxo-2,3-dihydro-5H-[1,3]thiazolo[3,2-a]pyridine-7-carboxylic acid - CTPC was elucidated using detailed NMR: one-dimensional (1H, 13C), as well as two-dimensional NMR spectra (1H-1H COSY, 1H-13C HSQC, 1H-13C HMBC, 1H-15N HSQC, 1H-15N HMBC) experiments. Besides, the essential optical, biological and antioxidative properties of CTPC were investigated.
Sigma-Aldrich
7-Diethylamino-4-methylcoumarin, 99%