MilliporeSigma
  • Use of supercritical carbon dioxide technology for fabricating a tissue engineering scaffold for anterior cruciate ligament repair.
Use of supercritical carbon dioxide technology for fabricating a tissue engineering scaffold for anterior cruciate ligament repair.
Scientific reports (2020-08-21)
Ines Sherifi, Manon Bachy, Thomas Laumonier, Hervé Petite, Didier Hannouche
ABSTRACT
Tissue-engineered grafts may be useful in Anterior Cruciate Ligament (ACL) repair and provide a novel, alternative treatment to clinical complications of rupture, harvest site morbidity and biocompatibility associated with autografts, allografts and synthetic grafts. We successfully used supercritical carbon dioxide (Sc-CO2) technology for manufacturing a "smart" biomaterial scaffold, which retains the native protein conformation and tensile strength of the natural ACL but is decellularized for a decreased immunogenic response. We designed and fabricated a new scaffold exhibiting (1) high tensile strength and biomechanical properties comparable to those of the native tissue, (2) thermodynamically-stable extra-cellular matrix (ECM), (3) preserved collagen composition and crosslinking, (4) a decellularized material milieu with potential for future engineering applications and (5) proven feasibility and biocompatibility in an animal model of ligament reconstruction. Because of the "smart" material ECM, this scaffold may have the potential for providing a niche and for directing stem cell growth, differentiations and function pertinent to new tissue formation. Sc-CO2-related technology is advanced and has the capability to provide scaffolds of high strength and durability, which sustain a lifetime of wear and tear under mechanical loading in vivo.
Sigma-Aldrich
Chloramine-T hydrate, 95%