MilliporeSigma
  • Novel apramycin resistance gene apmA in bovine and porcine methicillin-resistant Staphylococcus aureus ST398 isolates.
Novel apramycin resistance gene apmA in bovine and porcine methicillin-resistant Staphylococcus aureus ST398 isolates.
Antimicrobial agents and chemotherapy (2010-09-30)
Andrea T Fessler, Kristina Kadlec, Stefan Schwarz
ABSTRACT
A novel apramycin resistance gene, apmA, was detected on the ca.-40-kb resistance plasmid pAFS11 from bovine methicillin-resistant Staphylococcus aureus (MRSA) of sequence type 398 (ST398). The apmA gene coded for a protein of 274 amino acids that was related only distantly to acetyltransferases involved in chloramphenicol or streptogramin A resistance. NsiI deletion of apmA resulted in a 16- to 32-fold decrease in the apramycin MICs. An apmA-specific PCR identified this gene in one additional bovine and four porcine MRSA ST398 isolates.
Sigma-Aldrich
Apramycin sulfate salt
Sigma-Aldrich
Erythromycin, potency: ≥850 μg per mg
Sigma-Aldrich
Erythromycin, BioReagent, suitable for cell culture
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
Sigma-Aldrich
Erythromycin, meets USP testing specifications
Sigma-Aldrich
Erythromycin, tested according to Ph. Eur.
Sigma-Aldrich
Trimethoprim, ≥99.0% (HPLC)
Sigma-Aldrich
Trimethoprim, ≥98.5%
Supelco
Trimethoprim, VETRANAL®, analytical standard