- Lamellarin 14, a derivative of marine alkaloids, inhibits the T790M/C797S mutant epidermal growth factor receptor.
Lamellarin 14, a derivative of marine alkaloids, inhibits the T790M/C797S mutant epidermal growth factor receptor.
The emergence of acquired resistance is a major concern associated with molecularly targeted kinase inhibitors. The C797S mutation in the epidermal growth factor receptor (EGFR) confers resistance to osimertinib, a third-generation EGFR-tyrosine kinase inhibitor (EGFR-TKI). We report that the derivatization of the marine alkaloid topoisomerase inhibitor lamellarin N provides a structurally new class of EGFR-TKIs. One of these, lamellarin 14, is effective against the C797S mutant EGFR. Bioinformatic analyses revealed that the derivatization transformed the topoisomerase inhibitor-like biological activity of lamellarin N into kinase inhibitor-like activity. Ba/F3 and PC-9 cells expressing the EGFR in-frame deletion within exon 19 (del ex19)/T790M/C797S triple-mutant were sensitive to lamellarin 14 in a dose range similar to the effective dose for cells expressing EGFR del ex19 or del ex19/T790M. Lamellarin 14 decreased the autophosphorylation of EGFR and the downstream signaling in the triple-mutant EGFR PC-9 cells. Furthermore, intraperitoneal administration of 10 mg/kg lamellarin 14 for 17 days suppressed tumor growth of the triple-mutant EGFR PC-9 cells in a mouse xenograft model using BALB/c nu/nu mice. Thus, lamellarin 14 serves as a novel structural backbone for an EGFR-TKI that prevents the development of cross-resistance against known drugs in this class.