MilliporeSigma
  • The Impact of Aging, Psychotic Symptoms, Medication, and Brain-Derived Neurotrophic Factor on Cognitive Impairment in Japanese Chronic Schizophrenia Patients.
The Impact of Aging, Psychotic Symptoms, Medication, and Brain-Derived Neurotrophic Factor on Cognitive Impairment in Japanese Chronic Schizophrenia Patients.
Frontiers in psychiatry (2018-06-14)
Kiyokazu Atake, Tomoyuki Nakamura, Nobuhisa Ueda, Hikaru Hori, Asuka Katsuki, Reiji Yoshimura
ABSTRACT
Background: Cognitive impairment in schizophrenia can result in considerable difficulty in performing functions of daily life or social rehabilitation. Cognitive impairment in schizophrenia is related to various factors, such as the psychotic severity, aging, medication, and brain-derived neurotrophic factor (BDNF). To date, however, no studies investigating the impact of these factors on cognitive functioning in chronic schizophrenia patients have been performed. Objective: The aim of this study is to identify those factors that influence the cognitive functioning in patients with chronic schizophrenia. Methods: Sixty-five of 116 long-term hospitalized chronic schizophrenia patients (63.8 ± 12.1 years old, M/F = 29/36) were enrolled this cross-sectional study. We investigated the relationship among the patients' age, psychotic severity, treatment medication, serum BDNF levels, and cognitive functioning (measured by the Japanese-language version of the Brief Assessment of Cognition in Schizophrenia; BACS-J). Additionally, we performed a multivariable linear regression analysis. Results: According to the partial correlation analysis, certain parameters [i.e., age, chlorpromazine (CP) equivalent, biperiden (BP) equivalent, and serum BDNF] were significantly correlated with cognitive functioning, including working memory (WM), motor function (MF), attention and processing speed (AP), and executive function (EF). For the multivariate analysis, the MF component, which had the highest correlation, was selected as the dependent variable, and the independent variables included age, Manchester Scale for chronic psychosis (ManS) total score, CP equivalent, BP equivalent, serum BDNF, estimated full scale IQ, and years of education. According to the multiple regression analysis of this model, R (multiple regression coefficient) was 0.542, the adjusted R2 (coefficient of determination) was 0.201, and only BP equivalent (β = -0.305, p = 0.030), but not age, ManS score, CP equivalent, or serum BDNF, could significantly explain MF at the 5% significant level. Conclusion: In conclusion, aging, medication (administering more antipsychotics or anticholinergics), and serum BDNF concentration are significantly correlated with cognitive dysfunction in chronic schizophrenia patients but not with the severity of psychotic symptoms. Furthermore, only the anticholinergic dosage had a significant causal relationship with MF. Thus, the use of anticholinergics in chronic schizophrenia patients with deteriorating cognitive functioning must be reconsidered.
Product Description
Millipore
MILLIPLEX® Human Neurodegenerative Disease Panel