MilliporeSigma
  • Wnt4 antagonises Wnt3a mediated increases in growth and glucose stimulated insulin secretion in the pancreatic beta-cell line, INS-1.
Wnt4 antagonises Wnt3a mediated increases in growth and glucose stimulated insulin secretion in the pancreatic beta-cell line, INS-1.
Biochemical and biophysical research communications (2016-10-01)
A Bowen, K Kos, J Whatmore, S Richardson, H J Welters
ABSTRACT
The Wnt signalling pathway in beta-cells has been linked to the development of type 2 diabetes. Investigating the impact of a non-canonical Wnt ligand, Wnt4, on beta-cell function we found that in INS-1 cells, Wnt4 was able to completely block Wnt3a stimulated cell growth and insulin secretion. However, despite high levels of Wnt4 protein being detected in INS-1 cells, reducing the expression of Wnt4 had no impact on cell growth or Wnt3a signalling. As such, the role of the endogenously expressed Wnt4 in beta-cells is unclear, but the data showing that Wnt4 can act as a negative regulator of canonical Wnt signalling in beta-cells suggests that this pathway could be a potential target for modulating beta-cell function.
Sigma-Aldrich
Anti-β-Catenin Antibody, clone 2H4A7, clone 2H4A7, Upstate®, from mouse