MilliporeSigma
  • Less airway inflammation and goblet cell metaplasia in an IL-33-induced asthma model of leptin-deficient obese mice.
Less airway inflammation and goblet cell metaplasia in an IL-33-induced asthma model of leptin-deficient obese mice.
Respiratory research (2021-06-03)
Atsushi Kurokawa, Mitsuko Kondo, Ken Arimura, Shigeru Ashino, Etsuko Tagaya
ABSTRACT
Obesity-associated asthma is a phenotype of severe asthma. Late-onset, non-eosinophilic and female-dominant phenotype is highly symptomatic and difficult to treat. Leptin, an adipokine, exerts an immunomodulatory effect. IL-33 associated with innate immunity induces type 2 inflammation and is present in adipose tissue. The purpose of this study was to elucidate the pathogenesis of obesity-associated asthma by focusing on the interaction between leptin and IL-33. In leptin-deficient obese (ob/ob) and wild-type mice, IL-33 was instilled intranasally on three consecutive days. In part of the mice, leptin was injected intraperitoneally prior to IL-33 treatment. The mice were challenged with methacholine, and airway hyperresponsiveness (AHR) was assessed by resistance (Rrs) and elastance (Ers) of the respiratory system using the forced oscillation technique. Cell differentiation, IL-5, IL-13, eotaxin, keratinocyte-derived chemokine (KC) in bronchoalveolar lavage fluid (BALF) and histology of the lung were analyzed. For the in vitro study, NCI-H292 cells were stimulated with IL-33 in the presence or absence of leptin. Mucin-5AC (MUC5AC) levels were measured using an enzyme-linked immunosorbent assay. Ob/ob mice showed greater Rrs and Ers than wild-type mice. IL-33 with leptin, but not IL-33 alone, enhanced Ers rather than Rrs challenged with methacholine in ob/ob mice, whereas it enhanced Rrs alone in wild-type mice. IL-33-induced eosinophil numbers, cytokine levels in BALF, eosinophilic infiltration around the bronchi, and goblet cell metaplasia were less in ob/ob mice than in wild-type mice. However, leptin pretreatment attenuated these changes in ob/ob mice. MUC5AC levels were increased by co-stimulation with IL-33 and leptin in vitro. Ob/ob mice show innate AHR. IL-33 with leptin, but not IL-33 alone, induces airway inflammation and goblet cell metaplasia and enhances AHR involving peripheral airway closure. This is presumably accelerated by mucus in ob/ob mice. These results may explain some aspects of the pathogenesis of obesity-associated asthma.
Sigma-Aldrich
IL-33 from mouse, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture