Merck
  • PPP1CA/YAP/GS/Gln/mTORC1 pathway activates retinal Müller cells during diabetic retinopathy.
PPP1CA/YAP/GS/Gln/mTORC1 pathway activates retinal Müller cells during diabetic retinopathy.
Experimental eye research (2021-07-20)
Yang Guo, Xiaomin Cang, Linling Zhu, Manhui Zhu, Aihong Li, Zhenzhen Wang, Yuting Zhang, Xueqin Wang, E Song
ABSTRACT
Diabetic retinopathy (DR) is a vision-loss complication caused by diabetes with high prevalence. During DR, the retinal microvascular injury and neurodegeneration derived from chronic hyperglycemia have attracted global attention to retinal Müller cells (RMCs), the major macroglia in the retina contributes to neuroprotection. Protein Phosphatase 1 Catalytic Subunit Alpha (PPP1CA) dephosphorylates the transcriptional coactivator Yes-associated protein (YAP) to promote the transcription of glutamine synthetase (GS). GS catalyzes the transformation of neurotoxic glutamate (Glu) into nontoxic glutamine (Gln) to activate the mammalian target of rapamycin complex 1 (mTORC1), which promotes the activation of RMCs. In this study, in vitro MIO-M1 cell and in vivo mouse high-fat diet and streptozotocin (STZ)-induced diabetic model to explore the role of the PPP1CA/YAP/GS/Gln/mTORC1 pathway on the activation of MRCs during DR. Results showed that PPP1CA promoted the dephosphorylation and nuclear translocation of YAP in high glucose (HG)-exposed MIO-M1 cells. YAP transcribed GS in HG-exposed MIO-M1 cells in a TEAD1-dependent and PPP1CA-dependent way. GS promoted the biosynthesis of Gln in HG-exposed MIO-M1 cells. Gln activated mTORC1 instead of mTORC2 in HG-exposed MIO-M1 cells. The proliferation and activation of HG-exposed MIO-M1 cells were PPP1CA/YAP/GS/Gln/mTORC1-dependent. Finally, RMC proliferation and activation during DR were inhibited by the PPP1CA/YAP/GS/Gln/mTORC1 blockade. The findings supplied a potential idea to protect RMCs and alleviate the development of DR.
Sigma-Aldrich
D-2-Aminoadipic acid