Merck
  • Single-molecule fluorescence imaging to quantify membrane protein dynamics and oligomerization in living plant cells.
Single-molecule fluorescence imaging to quantify membrane protein dynamics and oligomerization in living plant cells.
Nature protocols (2015-11-20)
Xiaohua Wang, Xiaojuan Li, Xin Deng, Doan-Trung Luu, Christophe Maurel, Jinxing Lin
ABSTRACT
Measuring the mobility and interactions of proteins is key to understanding cellular signaling mechanisms; however, quantitative analysis of protein dynamics in living plant cells remains a major challenge. Here we describe an automated, single-molecule protocol based on total internal reflection fluorescence microscopy (TIRFM) imaging that allows protein tracking and subunit counting in living plant cells. This protocol uses TIRFM to image transgenic plant tissues expressing fluorescently tagged proteins that are localized to the plasma membrane. Next, a tracking algorithm quantifies dynamic changes in fluorescent protein motion types, temporary particle displacement and protein photobleaching steps. This protocol allows researchers to study the kinetic characteristics of heterogeneously distributed proteins. The approach has potential applications for studies of protein dynamics and subunit stoichiometry for a wide variety of plasma membrane and intracellular proteins in living plant cells and other biological specimens visualized by TIRFM or other fluorescence imaging techniques. The whole protocol can be completed in 5-6 h.
Sigma-Aldrich
Agar, Type M, suitable for plant cell culture