MilliporeSigma
  • Photoaffinity labelling of carnitine acetyltransferase with S-(p-azidophenacyl)thiocarnitine.
Photoaffinity labelling of carnitine acetyltransferase with S-(p-azidophenacyl)thiocarnitine.
The Biochemical journal (1986-07-15)
J M Mauro, R V Lewis, R E Barden
ABSTRACT
A photolabile reagent, p-azidophenacyl-DL-thiocarnitine, was synthesized and tested as a photoaffinity label for carnitine acetyltransferase (EC 2.3.1.7) from pigeon breast. p-Azidophenacyl-DL-thiocarnitine is an active-site-directed reagent for this acetyltransferase, since it is a competitive inhibitor (Ki 10 microM) versus carnitine. U.v. irradiation of a mixture of p-azidophenacyl-DL-thiocarnitine and enzyme produces irreversible inhibition. Acetyl-DL-carnitine protects the enzyme from inhibition by photoactivated p-azidophenacyl-DL-thiocarnitine. In the presence of 30 mM-2-mercaptoethanol as a scavenger, the relationship between loss of activity and photoincorporation of reagent suggests that one molecule of reagent is incorporated per molecule of inhibited enzyme. However, peptide maps of enzyme labelled with p-azidophenacyl[14C]thiocarnitine indicate that several (about six) tryptic peptides (of a possible 60-65) are modified. The presence of 5 mM-acetyl-DL-carnitine significantly decreases the incorporation of reagent in each labelled tryptic peptide.
Sigma-Aldrich
Acetyl-DL-carnitine hydrochloride