Merck
  • Paradoxical effects of green tea (Camellia sinensis) and antioxidant vitamins in diabetic rats: improved retinopathy and renal mitochondrial defects but deterioration of collagen matrix glycoxidation and cross-linking.
Paradoxical effects of green tea (Camellia sinensis) and antioxidant vitamins in diabetic rats: improved retinopathy and renal mitochondrial defects but deterioration of collagen matrix glycoxidation and cross-linking.
Diabetes (2005-01-29)
Georgian T Mustata, Mariana Rosca, Klaus M Biemel, Oliver Reihl, Mark A Smith, Ashwini Viswanathan, Christopher Strauch, Yunpeng Du, Jie Tang, Timothy S Kern, Markus O Lederer, Michael Brownlee, Miriam F Weiss, Vincent M Monnier
ABSTRACT
We tested the hypothesis that green tea prevents diabetes-related tissue dysfunctions attributable to oxidation. Diabetic rats were treated daily with tap water, vitamins C and E, or fresh Japanese green tea extract. After 12 months, body weights were decreased, whereas glycated lysine in aorta, tendon, and plasma were increased by diabetes (P < 0.001) but unaffected by treatment. Erythrocyte glutathione and plasma hydroperoxides were improved by the vitamins (P < 0.05) and green tea (P < 0.001). Retinal superoxide production, acellular capillaries, and pericyte ghosts were increased by diabetes (P < 0.001) and improved by green tea and the vitamins (P variable). Lens crystallin fluorescence at 370/440 nm was ameliorated by green tea (P < 0.05) but not the vitamins. Marginal effects on nephropathy parameters were noted. However, suppressed renal mitochondrial NADH-linked ADP-dependent and dinitrophenol-dependent respiration and complex III activity were improved by green tea (P variable). Green tea also suppressed the methylglyoxal hydroimidazolone immunostaining of a 28-kDa mitochondrial protein. Surprising, glycoxidation in tendon, aorta, and plasma was either worsened or not significantly improved by the vitamins and green tea. Glucosepane cross-links were increased by diabetes (P < 0.001), and green tea worsened total cross-linking. In conclusion, green tea and antioxidant vitamins improved several diabetes-related cellular dysfunctions but worsened matrix glycoxidation in selected tissues, suggesting that antioxidant treatment tilts the balance from oxidative to carbonyl stress in the extracellular compartment.
Sigma-Aldrich
Glutathione Assay Kit