MilliporeSigma
  • Modulation of the tick gut milieu by a secreted tick protein favors Borrelia burgdorferi colonization.
Modulation of the tick gut milieu by a secreted tick protein favors Borrelia burgdorferi colonization.
Nature communications (2017-08-05)
Sukanya Narasimhan, Tim J Schuijt, Nabil M Abraham, Nallakkandi Rajeevan, Jeroen Coumou, Morven Graham, Andrew Robson, Ming-Jie Wu, Sirlei Daffre, Joppe W Hovius, Erol Fikrig
ABSTRACT
The Lyme disease agent, Borrelia burgdorferi, colonizes the gut of the tick Ixodes scapularis, which transmits the pathogen to vertebrate hosts including humans. Here we show that B. burgdorferi colonization increases the expression of several tick gut genes including pixr, encoding a secreted gut protein with a Reeler domain. RNA interference-mediated silencing of pixr, or immunity against PIXR in mice, impairs the ability of B. burgdorferi to colonize the tick gut. PIXR inhibits bacterial biofilm formation in vitro and in vivo. Abrogation of PIXR function in vivo results in alterations in the gut microbiome, metabolome and immune responses. These alterations influence the spirochete entering the tick gut in multiple ways. PIXR abrogation also impairs larval molting, indicative of its role in tick biology. This study highlights the role of the tick gut in actively managing its microbiome, and how this impacts B. burgdorferi colonization of its arthropod vector. Borrelia burgdorferi, the causative agent of Lyme disease, is transmitted by the tick Ixodes scapularis. Here, the authors show that a tick secreted protein (PIXR) modulates the tick gut microbiota and facilitates B. burgdorferi colonization.
Sigma-Aldrich
Glycoprotein Detection Kit, Sufficient for 10 mini-gels (10 × 10 cm) or 5 gels or membranes (16.5 × 19 cm)