Merck
  • Evaluation of Angiogenesis-Related Genes as Prognostic Biomarkers of Bevacizumab Treated Ovarian Cancer Patients: Results from the Phase IV MITO16A/ManGO OV-2 Translational Study.
Evaluation of Angiogenesis-Related Genes as Prognostic Biomarkers of Bevacizumab Treated Ovarian Cancer Patients: Results from the Phase IV MITO16A/ManGO OV-2 Translational Study.
Cancers (2021-10-24)
Daniela Califano, Daniela Gallo, Gian Luca Rampioni Vinciguerra, Rossella De Cecio, Laura Arenare, Simona Signoriello, Daniela Russo, Gabriella Ferrandina, Francesca Citron, Nunzia Simona Losito, Piera Gargiulo, Vittorio Simeon, Giovanni Scambia, Sabrina Chiara Cecere, Marco Montella, Nicoletta Colombo, Germana Tognon, Eliana Bignotti, Gian Franco Zannoni, Vincenzo Canzonieri, Alessandra Ciucci, Anna Spina, Giosuè Scognamiglio, Michele Del Sesto, Clorinda Schettino, Maria Carmela Piccirillo, Francesco Perrone, Paolo Chiodini, Sandro Pignata, Gustavo Baldassarre
ABSTRACT
Epithelial ovarian cancer (EOC) is a rare, highly lethal disease. In a subset of high grade EOC patients, maintenance therapy with the antiangiogenic drug Bevacizumab (BEV) is a valuable option. To date, no validated predictive or prognostic biomarkers exist for selecting EOC patients that might benefit from BEV treatment. Immunohistochemistry and RT-qPCR evaluated the expression of seven angiogenesis-related proteins and of a twelve microRNAs angio-signature in EOC patients, treated in first line with chemotherapy plus BEV (MITO16A/ManGO OV-2 phase IV trial). Centralized statistical analyses assessed the associations between each biomarker, clinical prognostic factors and survival outcomes. High miR-484 expression was associated with longer progression-free and overall survival. Notably, the combined expression of miR-484 and its target VEGFB identified a subset of patients that might mostly benefit from BEV treatment. No other significant correlations were found between the other analyzed biomarkers and patients' survival. The application of a shrinkage procedure to adjust for over-fitting hazard ratio estimates reduced the association significance. The analysis of angiogenesis related biomarkers in EOC patients homogenously treated with BEV in first line provides novel insight in their prognostic value and suggests that some of them might merit to be tested as predictive markers of drug activity in dedicated randomized trials.
Sigma-Aldrich
Monoclonal Anti-FLT1 antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL0344, purified immunoglobulin, buffered aqueous glycerol solution