MilliporeSigma
  • LINC00958 promotes bladder cancer carcinogenesis by targeting miR-490-3p and AURKA.
LINC00958 promotes bladder cancer carcinogenesis by targeting miR-490-3p and AURKA.
BMC cancer (2021-10-28)
Hongtao Zhen, Peng Du, Qiang Yi, Xiaolong Tang, Tongqing Wang
ABSTRACT
Bladder cancer is a prevalent malignancy of the urinary system, in which long non-coding RNAs (lncRNAs) are highly associated. We aimed to elucidate the role of LINC00958 in bladder cancer. LINC00958 expression levels were measured using qRT-PCR. The interaction of LINC00958-miR-490-3p-AURKA was analyzed by luciferase, RIP, and RNA pull-down assays. The biological roles of LINC00958, miR-490-3p, and AURKA in bladder cancer cells were analyzed using CCK8, BrdU, and transwell assays. Increased expression of LINC00958 and AURKA was observed in bladder cancer tissues and cell lines. Decreased LINC00958 expression repressed bladder cancer progression and downregulation of miR-490-3p accelerated bladder cancer cell progression. Moreover, LINC00958 sponges miR-490-3p to upregulate AURKA expression, thereby promoting carcinogenesis in bladder cancer cells. Our study revealed that LINC00958 facilitated cell proliferation and invasion, and suppressed cell apoptosis by sponging miR-490-3p and upregulating AURKA, thus inspiring a new treatment method for bladder cancer.
Product Description
Millipore
RIPA Lysis Buffer, 10X, 100 mL RIPA Lysis Buffer, 10X for Immunoprecipitation & Western Blotting.
Sigma-Aldrich
Crystal Violet, certified by the Biological Stain Commission
Sigma-Aldrich
Magna RIP® RNA-Binding Protein Immunoprecipitation Kit, RNA Immunoprecipitation (RIP) Kit containing all necessary reagents to perform 12 individual RNA-binding protein immunoprecipitation (RIP) reactions using protein A/G magnetic beads.