Merck
  • X-Linked TLR7 Deficiency Underlies Critical COVID-19 Pneumonia in a Male Patient with Ataxia-Telangiectasia.
X-Linked TLR7 Deficiency Underlies Critical COVID-19 Pneumonia in a Male Patient with Ataxia-Telangiectasia.
Journal of clinical immunology (2021-10-24)
Hassan Abolhassani, Ahmad Vosughimotlagh, Takaki Asano, Nils Landegren, Bertrand Boisson, Samaneh Delavari, Paul Bastard, Maribel Aranda-Guillén, Yating Wang, Fanglei Zuo, Fabian Sardh, Harold Marcotte, Likun Du, Shen-Ying Zhang, Qian Zhang, Nima Rezaei, Olle Kämpe, Jean-Laurent Casanova, Lennart Hammarström, Qiang Pan-Hammarström
ABSTRACT
Coronavirus disease 2019 (COVID-19) exhibits a wide spectrum of clinical manifestations, ranging from asymptomatic to critical conditions. Understanding the mechanism underlying life-threatening COVID-19 is instrumental for disease prevention and treatment in individuals with a high risk. We aimed to identify the genetic cause for critical COVID-19 pneumonia in a patient with a preexisting inborn error of immunity (IEI). Serum levels of specific antibodies against the virus and autoantibodies against type I interferons (IFNs) were measured. Whole exome sequencing was performed, and the impacts of candidate gene variants were investigated. We also evaluated 247 ataxia-telangiectasia (A-T) patients in the Iranian IEI registry. We report a 7-year-old Iranian boy with a preexisting hyper IgM syndrome who developed critical COVID-19 pneumonia. IgM only specific COVID-19 immune response was detected but no autoantibodies against type I IFN were observed. A homozygous deleterious mutation in the ATM gene was identified, which together with his antibody deficiency, radiosensitivity, and neurological signs, established a diagnosis of A-T. Among the 247 A-T patients evaluated, 36 had SARS-CoV-2 infection, but all had mild symptoms or were asymptomatic except the index patient. A hemizygous deleterious mutation in the TLR7 gene was subsequently identified in the patient. We report a unique IEI patient with combined ATM and TLR7 deficiencies. The two genetic defects underlie A-T and critical COVID-19 in this patient, respectively.
Sigma-Aldrich
IFN-omega human, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture