MilliporeSigma
  • Hereditary pancreatitis is caused by a mutation in the cationic trypsinogen gene.
Hereditary pancreatitis is caused by a mutation in the cationic trypsinogen gene.
Nature genetics (1996-10-01)
D C Whitcomb, M C Gorry, R A Preston, W Furey, M J Sossenheimer, C D Ulrich, S P Martin, L K Gates, S T Amann, P P Toskes, R Liddle, K McGrath, G Uomo, J C Post, G D Ehrlich
ABSTRACT
Hereditary pancreatitis (HP) is a rare, early-onset genetic disorder characterized by epigastric pain and often more serious complications. We now report that an Arg-His substitution at residue 117 of the cationic trypsinogen gene is associated with the HP phenotype. This mutation was observed in all HP affected individuals and obligate carriers from five kindreds, but not in individuals who married into the families nor in 140 unrelated individuals. X-ray crystal structure analysis, molecular modelling, and protein digest data indicate that the Arg 117 residue is a trypsin-sensitive site. Cleavage at this site is probably part of a fail-safe mechanism by which trypsin, which is activated within the pancreas, may be inactivated; loss of this cleavage site would permit autodigestion resulting in pancreatitis.
Sigma-Aldrich
Trypsinogen, PMSF treated from bovine pancreas, For use as a marker in SDS-PAGE