- Genome-scale deconvolution of RNA structure ensembles.
Genome-scale deconvolution of RNA structure ensembles.
Nature methods (2021-02-24)
Edoardo Morandi, Ilaria Manfredonia, Lisa M Simon, Francesca Anselmi, Martijn J van Hemert, Salvatore Oliviero, Danny Incarnato
PMID33619392
ABSTRACT
RNA structure heterogeneity is a major challenge when querying RNA structures with chemical probing. We introduce DRACO, an algorithm for the deconvolution of coexisting RNA conformations from mutational profiling experiments. Analysis of the SARS-CoV-2 genome using dimethyl sulfate mutational profiling with sequencing (DMS-MaPseq) and DRACO, identifies multiple regions that fold into two mutually exclusive conformations, including a conserved structural switch in the 3' untranslated region. This work may open the way to dissecting the heterogeneity of the RNA structurome.