MilliporeSigma
  • Generation and persistence of S1 IgG and neutralizing antibodies in post-COVID-19 patients.
Generation and persistence of S1 IgG and neutralizing antibodies in post-COVID-19 patients.
Infection (2021-10-21)
Oscar Guzmán-Martínez, Kathia Guardado, Miguel Varela-Cardoso, Alejandro Trujillo-Rivera, Antonio Marín-Hernández, María Cristina Ortiz-León, Iván Gómez-Ñañez, Michelle Gutiérrez, Rafaela Espinosa, Clara Luz Sampieri, Celso Ramos, Delia Vanesa López-Guerrero, Roberto Zenteno-Cuevas, Julio Isael Pérez-Carreón, Guadalupe Canales-Vázquez, Hilda Montero
ABSTRACT
Antibody-mediated immune response plays an important role in protection against reinfection. In the case of SARS-CoV-2 infection, the maximum duration of antibody response is still unknown. In this work, the generation of neutralizing  antibodies (NAbs) and IgG antibodies against the S1 subunit (S1 IgG ) of SARS-CoV-2 and their possible duration were determined through decay models. 132 participants with SARS-CoV-2 infection were classified according to the severity of the disease. Seroconversion and persistence of S1 IgG antibodies and NAbs were determined by ELISA, samples were taken at two different times post-infection and duration of those antibodies was estimated using Linear Mixed Models (LMMs). The highest amount of S1 IgGs antibodies was associated with age (41 years or older), greater severity of COVID-19 and male gender. NAbs production was associated with the same variables, except for age. The percentage of NAbs decay is higher in the asymptomatic group (P = 0.033), while in S1 IgG antibodies decay, no statistical difference was found between the 4 severity groups. An exponential decay model was built by using a LMM and similarly, two dispersion regions where constructed. The duration of S1 IgG antibodies was 744 days (668-781) for first region and 744 days (453-1231) for the second. Regarding NAbs, an adaptative LMM was used to model a logistic function, determining a duration of 267 days (215-347). Humoral immunity to SARS-CoV-2 infection depends on the severity of the disease, gender and age. This immune response could be long-lasting as for other coronaviruses.
Sigma-Aldrich
SIGMAFAST OPD, tablet