MilliporeSigma
  • Biomass productivity and productivity of fatty acids and amino acids of microalgae strains as key characteristics of suitability for biodiesel production.
Biomass productivity and productivity of fatty acids and amino acids of microalgae strains as key characteristics of suitability for biodiesel production.
Journal of applied phycology (2012-11-06)
Niels Hempel, Ingolf Petrick, Frank Behrendt
ABSTRACT
Microalgae are discussed as an alternative source for the production of biofuels. The lipid content compared to cultivation time of used species is the main reason for any choice of a special strain. This paper reviews more analytical data of 38 screened microalgae strains. After the cultivation period, total content of lipids was analysed. The extracted fatty acids were quantified as fatty acid methyl esters by GC analysis. The amino acids were analysed by HPLC. Chlorella sp., Chlorella saccharophila, Chlorella minutissima and Chlorella vulgaris were identified as species with the highest productivity of fatty acids relevant to transesterification reactions. The components were mainly linoleic acid, palmitic acid and oleic acid. To increase productivity of highly saturated fatty acids, cultivation parameters light intensity and temperature were varied. In this manner, the ideal conditions for biodiesel production were defined in this publication. ELECTRONIC SUPPLEMENTARY MATERIAL: The online version of this article (doi:10.1007/s10811-012-9795-3) contains supplementary material, which is available to authorized users.
Product Description
Supelco
4-(Dimethylamino)benzaldehyde solution, for TLC derivatization, 10 g/L in isopropanol