MilliporeSigma
Intestinal IgA synthesis: regulation of front-line body defences.
Nature reviews. Immunology (2003-01-04)
Sidonia Fagarasan, Tasuku Honjo
ABSTRACT
Immunoglobulin A is the most abundant immunoglobulin isotype in mucosal secretions. In this review, we summarize recent advances in our understanding of the sites, mechanisms and functions of intestinal IgA synthesis in mice. On the basis of these recent findings, we propose an updated model for the induction and regulation of IgA responses in the gut. In addition, we discuss new insights into the role of IgA in the maintenance of gut homeostasis and into the reciprocal interactions between gut B cells and the bacterial flora.