MilliporeSigma
  • Determination of Mogroside V in Luohanguo Extract for Daily Quality Control Operation Using Relative Molar Sensitivity to Single-Reference Caffeine.
Determination of Mogroside V in Luohanguo Extract for Daily Quality Control Operation Using Relative Molar Sensitivity to Single-Reference Caffeine.
Chemical & pharmaceutical bulletin (2021-01-05)
Naoko Masumoto, Kyoko Ishizuki, Yuzo Nishizaki, Takashi Ohtsuki, Miho Kuroe, Taichi Yamazaki, Masahiko Numata, Hiroshi Matsufuji, Naoki Sugimoto, Kyoko Sato
ABSTRACT
Mogroside V is one of the characteristic and effective components of luohanguo extract, a food additive used as a sweetener in Japan as per Japan's Standards and Specifications for Food Additives (JSFA; 9th ed.). JSFA stipulates that the quantitative determination for mogroside V content in luohanguo extract applies HPLC using analytical standard mogroside V. However, no mogroside V reagents with proven purities are commercially available. Therefore the current JSFA determination method is not particularly suited for daily quality control operations involving luohanguo extract. In this study, we applied an alternative quantitative method using a single reference with relative molar sensitivity (RMS). It was possible to calculate the accurate RMS by an offline combination of 1H-quantitative NMR spectroscopy (1H-qNMR) and an HPLC/variable-wavelength detector (VWD). Using the RMS of mogroside V to a commercial certified reference material grade caffeine, the mogroside V contents in luohanguo extracts could be determined using HPLC/VWD without analytical standard mogroside V. There was no significant difference between the mogroside V contents in luohanguo extracts determined using the method employing single-reference caffeine with the RMS and using the JSFA method. The absolute calibration curve for the latter was prepared using an analytical standard mogroside V whose purity was determined by 1H-qNMR. These results demonstrate that our proposed method using a single reference with RMS is suitable for quantitative determination of mogroside V in luohanguo extract and can be used as an alternative method to the current assay method in JSFA.
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D
Supelco
Caffeine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland