MilliporeSigma
  • The Impact of the CD9 Tetraspanin on Lentivirus Infectivity and Exosome Secretion.
The Impact of the CD9 Tetraspanin on Lentivirus Infectivity and Exosome Secretion.
Molecular therapy : the journal of the American Society of Gene Therapy (2017-12-10)
Kai O Böker, Nicolas Lemus-Diaz, Rafael Rinaldi Ferreira, Lara Schiller, Stefan Schneider, Jens Gruber
ABSTRACT
Efficient transduction tools are a hallmark for both research and therapy development. Here, we introduce new insights into the generation of lentiviral vectors with improved performance by utilizing producer cells with increased production rates of extracellular vesicles through CD9 overexpression. Most human cells secrete small vesicles from their surface (microvesicles) or intraluminal endosome-derived membranes (exosomes). In particular, enhanced levels of the tetraspanin CD9 result in significantly increased numbers of extracellular vesicles with exosome-like features that were secreted from four different human cell lines. Intriguingly, exosomes and their biogenesis route display similarities to lentivirus and we examined the impact of CD9 expression on release and infectivity of recombinant lentiviral vectors. Although the titers of released viral particles were not increased upon production in high CD9 cells, we observed improved performance in terms of both speed and efficiency of lentiviral gene delivery into numerous human cell lines, including HEK293, HeLa, SH-SY5Y, as well as B and T lymphocytes. Here, we demonstrate that enhanced CD9 enables lentiviral transduction in the absence of any pseudotyping viral glycoprotein or fusogenic molecule. Our findings indicate an important role of CD9 for lentiviral vector and exosome biogenesis and point out a remarkable function of this tetraspanin in membrane fusion, viral infectivity, and exosome-mediated horizontal information transfer.
Product Description
Supelco
Poly(ethyleneimine) solution, analytical standard, 50 % (w/v) in H2O
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse