MilliporeSigma
  • Evaluation of human antibody responses to keyhole limpet hemocyanin on a carbohydrate microarray.
Evaluation of human antibody responses to keyhole limpet hemocyanin on a carbohydrate microarray.
Proteomics. Clinical applications (2010-12-08)
Oyindasola Oyelaran, Jeffrey C Gildersleeve
ABSTRACT
Keyhole limpet hemocyanin (KLH) is used as a vaccine adjuvant, as a carrier protein for small haptens, and as a treatment for bladder cancer. Immunization with KLH produces antibodies to tumor-associated carbohydrate antigens (TACAs) in animals, and these antibodies have been postulated as the basis of efficacy for bladder cancer treatment. The purpose of this study was to evaluate antibody responses to KLH in humans. A carbohydrate microarray was used to profile antibody responses in 14 individuals immunized with KLH plus alum adjuvant. Eight out of fourteen individuals produced antibodies to at least one TACA. Increases to Lewis X, Lewis Y, GA1di, GM3, and sialyl Lewis A were observed in certain individuals, but, in general, antibody profiles were highly variable. Pre-immunization antibody levels to a subset of array antigens had a statistically significant correlation with the magnitude of the antibody response to KLH. Antibodies to TACAs can be produced in humans, but antibody profiles differ considerably from person to person, which may contribute to variable clinical responses with KLH. Pre-treatment antibody levels to certain antigens may be useful for predicting which patients will respond favorably to KLH.
Sigma-Aldrich
Hemocyanin, Keyhole Limpet, Megathura crenulata, High Purity, Endotoxin-Free, Sterile-Liquid, Hemocyanin, Keyhole Limpet, Megathura crenulata, High Purity, Endotoxin Free, is a large, multi-subunit, O2-carrying, metalloprotein that is used as a carrier protein in the production of antibodies.
Sigma-Aldrich
Hemocyanin, Keyhole Limpet, Megathura crenulata, BES/Mg2+ Buffer, Hemocyanin, Keyhole Limpet, Megathura crenulata, BES/Mg2+ buffer, CAS 9013-72-3, is a large, multi-subunit, O2-carrying, metalloprotein that is used in the production of antibodies.