MilliporeSigma
  • A novel LMNA indel mutation identified in a family with atrioventricular block and atrial fibrillation.
A novel LMNA indel mutation identified in a family with atrioventricular block and atrial fibrillation.
Medicine (2021-06-10)
Zhangrong Jia, Yue'an Zhang, Junping Deng, Yanqing Guo, Yimei Du, Gang Wang, Jiyao Xu, Xiaoming Li
ABSTRACT
It is well known that many genetic factors are involved in the occurrence and progression of atrioventricular block (AV block) and atrial fibrillation (AF). However, the genetic variants discovered so far have only explained parts of these processes. More genes and variants remain to be identified. In the present study, a three-generation family with an autosomal dominant form of AV block and AF was enrolled. Whole exome sequencing was conducted in three affected and one unaffected family member. A total of 64 nonsynonymous variants was shared by three affected individuals and not present in the unaffected individual. By selection of variants absent in the known databases and were predicted to be deleterious, 4 novel variants were identified. Only one novel frameshift insertion in the LMNA gene (c.825_826insCAGG) was identified in another affected family member and not detected in other non-affected family members and the 100 controls. Our finding expanded the spectrum of variants associated with AV block and AF, and was valuable in the genetic diagnosis of AV block and AF.