MilliporeSigma
  • Protective effect of neurotensin receptor-1 agonist PD 149163 against lipopolysaccharide-induced gut toxicity in mice.
Protective effect of neurotensin receptor-1 agonist PD 149163 against lipopolysaccharide-induced gut toxicity in mice.
Drug and chemical toxicology (2021-08-03)
Ankit Mishra, K P Singh
ABSTRACT
The interaction between neuroendocrine and immune components of the gut maintains the organism's physical and psychological health. Its disruption may reflect in disease conditions such as inflammatory bowel disease (IBD) and mental illness. The lipopolysaccharide (LPS) disrupts the endocrine-immune homeostasis resulting in gut toxicity. The Neurotensin receptor-1 (NTR-1) agonist PD 149163 (PD) acts as an atypical antipsychotic drug in psychiatric illness, but its role in modulating gut pathophysiology remains unknown. Therefore, the aim of the present study was to evaluate the protective effect of PD against LPS-induced gut toxicity. Swiss albino female mice (12 weeks) were divided into six groups (n = 6/group): (I) Control, (II) LPS (1 mg/kg, for 5 days), (III) LPS (1 mg/kg, for 5 days)+PD low (100 µg/kg, for 21 days), (IV) LPS (1 mg/kg, for 5 days)+PD high (300 µg/kg, for 21 days), (V) PD low (100 µg/kg, for 21 days), and (VI) PD high (300 µg/kg, for 21 days). Drugs were given intraperitoneal in the morning. PD administration prevented the LPS-induced gut inflammation observed in damage of epithelial barrier, disruption of goblet cells, and condensation of lamina propria (LP). The LPS-induced oxidative stress characterized by decreased superoxide dismutase (SOD) activity and increased lipid hydroperoxide (LOOH) (p < 0.001 for both), and enhanced interleukine-6 (IL-6) & tumor necrosis factor-α (TNF-α) (p < 0.001 for both) as well as immunointensity of NT (p < 0.01) and NTR-1 (p < 0.05) were reversed and normalized to control after PD treatment. Thus, the anti-inflammatory, anti-oxidative, and cell proliferation properties of PD modulate the gut toxicity in LPS-challenged mice.