MilliporeSigma
  • Endothelial nitric oxide synthase is expressed in amacrine cells of developing human retinas.
Endothelial nitric oxide synthase is expressed in amacrine cells of developing human retinas.
Investigative ophthalmology & visual science (2006-04-28)
Shengxiu Li, David Tay, Siyun Shu, Xinmin Bao, Yongming Wu, Xiaoyang Wang, Henry K Yip
ABSTRACT
To examine the expression and cellular distribution pattern of endothelial nitric oxide synthase (eNOS) in the developing human retina and to compare its expression with that in rats. Expression of eNOS was examined by immunohistochemistry in retinas of humans ranging from 8.5 to 28 weeks of gestation (WG) and of rats. In the developing human retina, eNOS expression was first detected in the proximal margin of the neuroblastic layer in the incipient fovea-surrounding area at 12 WG. At 17 to 28 WG, eNOS-immunoreactive cells were located in the innermost part of the inner nuclear layer and in the ganglion cell layer, expanding to both temporal and nasal retinas and the processes projecting into the inner plexiform layer. These eNOS-positive cells coexpressed syntaxin and glutamate decarboxylase, and are probably GABAergic amacrine cells. The onset of eNOS expression in developing amacrine cells, however, preceded the invasion of retinal vasculature, long before vascular function involving these cells can be expected, suggesting that eNOS has a role not only in vasoregulation but also in retinal development. From 20 WG on, eNOS was also detected in the photoreceptors adjacent to the fovea. eNOS expression in amacrine cells and photoreceptors was observed in the central-to-peripheral and temporal-to-nasal gradients. However, in the developing rat retina, eNOS was expressed exclusively in the vascular endothelial cells. The results support that eNOS plays a role, not only in the regulation of vascular function but also in the process of retinal development in humans.