MilliporeSigma
  • Phosphodiesterase 4D promotes angiotensin II-induced hypertension in mice via smooth muscle cell contraction.
Phosphodiesterase 4D promotes angiotensin II-induced hypertension in mice via smooth muscle cell contraction.
Communications biology (2022-01-22)
Tianfei Fan, Yangfeng Hou, Weipeng Ge, Tianhui Fan, Xiaohang Feng, Wenjun Guo, Xiaomin Song, Ran Gao, Jing Wang
ABSTRACT
Hypertension is a common chronic disease, which leads to cardio-cerebrovascular diseases, and its prevalence is increasing. The cyclic adenosine monophosphate (cAMP)-protein kinase A (PKA) pathway participates in multiple cardiovascular diseases. Phosphodiesterase (PDE) 4 has been shown to regulate PKA activity via cAMP specific hydrolysis. However, whether PDE4-cAMP-PKA pathway influences hypertension remains unknown. Herein, we reveal that PDE4D (one of PDE4 isoforms) expression is upregulated in the aortas of experimental hypertension induced by angiotensin II (Ang II). Furthermore, knockout of Pde4d in mouse smooth muscle cells (SMCs) attenuates Ang II-induced hypertension, arterial wall media thickening, vascular fibrosis and vasocontraction. Additionally, we find that PDE4D deficiency activates PKA-AMP-activated protein kinase (AMPK) signaling pathway to inhibit myosin phosphatase targeting subunit 1 (MYPT1)-myosin light chain (MLC) phosphorylation, relieving Ang II-induced SMC contraction in vitro and in vivo. Our results also indicate that rolipram, a PDE4 inhibitor, may be a potential drug for hypertension therapy.
Sigma-Aldrich
Rolipram, solid, ≥98% (HPLC)