MilliporeSigma
  • Quantitative Analysis of Cargo Density in Single-extracellular Vesicles by Imaging.
Quantitative Analysis of Cargo Density in Single-extracellular Vesicles by Imaging.
Bio-protocol (2018-12-20)
Taketoshi Kajimoto, Shun-Ichi Nakamura
ABSTRACT
Function of extracellular vesicles such as exosomes and microvesicles is determined by their wide ranges of cargoes inside them. Even in the pure exosomes or microvesicles the cargo contents are very heterogeneous. To understand this heterogeneous nature of extracellular vesicles, we need information of the vesicles, which will give us some parameters including vesicle size, number and cargo content of each vesicle. Here, we describe a new method to quantify cargo density in single-extracellular vesicles. Staining of extracellular vesicles in a membrane lipid content-proportionate manner and immobilization of extracellular vesicles onto glass substrate allow us to obtain cargo density information of single-extracellular vesicles. This protocol will be useful to analyze the effects of various drugs or genetic manipulation on vesicle generation and maturation including cargo sorting into heterogeneous extracellular vesicles.
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, pH 5.2, ≥96%
Sigma-Aldrich
Albumin, biotin labeled bovine, lyophilized powder