Merck
  • Base-catalyzed C-alkylation of potassium enolates with styrenes via a metal-ene reaction: a mechanistic study.
Base-catalyzed C-alkylation of potassium enolates with styrenes via a metal-ene reaction: a mechanistic study.
Organic & biomolecular chemistry (2020-02-27)
Joshua P Barham, Thierry N J Fouquet, Yasuo Norikane
ABSTRACT
Base-catalyzed, C-alkylation of potassium (K) enolates with styrenes (CAKES) has recently emerged as a highly practical and convenient method for elaboration or synthesis of pharmaceutically-relevant cores. K enolate-type precursors such as alkyl-substituted heterocycles (pyridines, pyrazines and thiophenes), ketones, imines, nitriles and amides undergo C-alkylation reactions with styrene in the presence of KOtBu or KHMDS. Surprisingly, no studies have probed the reaction mechanism beyond the likely initial formation of a K enolate. Herein, a synergistic approach of computational (DFT), kinetic and deuterium labelling studies rationalizes various experimental observations and supports a metal-ene-type reaction for amide CAKES. Moreover, our approach explains experimental observations in other reported C-alkylation reactions of other enolate-type precursors, thus implicating a general mechanism for CAKES.
Sigma-Aldrich
N-Methyl-d3-2-pyrrolidinone-d6, 98 atom % D