MilliporeSigma
  • Alterations in extracellular tryptophan and dopamine concentrations in rat striatum following peripheral administration of D- and L-tryptophan: an in vivo microdialysis study.
Alterations in extracellular tryptophan and dopamine concentrations in rat striatum following peripheral administration of D- and L-tryptophan: an in vivo microdialysis study.
Neuroscience letters (2012-08-14)
Shunsuke Yoshihara, Hayato Otani, Makoto Tsunoda, Kana Ishii, Hideaki Iizuka, Hideaki Ichiba, Takeshi Fukushima
ABSTRACT
Incorporation profiles of d-Trp and l-Trp into the striatum following intraperitoneal (i.p.) administration of d-Trp or l-Trp in male Sprague-Dawley rats (100mg/kg) were investigated by using a brain microdialysis technique. Alterations in the extracellular dopamine (DA) concentration in the rat striatum were also examined. Incorporation profiles of d-Trp and l-Trp were almost identical; however, transient DA release was only observed 0-30 min following d-Trp administration. Pretreatment with 3-methylpyrazole-5-carboxylic acid, an inhibitor of d-amino acid oxidase (DAAO), significantly suppressed the DA release induced by d-Trp. These findings suggest that d-Trp-induced DA release may be mediated by certain d-Trp metabolites produced by DAAO.
Sigma-Aldrich
D-Amino Acid Oxidase from porcine kidney, powder, ≥1.5 units/mg solid