Merck
  • Nitrogen nutrition contributes to plant fertility by affecting meiosis initiation.
Nitrogen nutrition contributes to plant fertility by affecting meiosis initiation.
Nature communications (2022-01-27)
Han Yang, Yafei Li, Yiwei Cao, Wenqing Shi, En Xie, Na Mu, Guijie Du, Yi Shen, Ding Tang, Zhukuan Cheng
ABSTRACT
Nitrogen (N), one of the most important plant nutrients, plays crucial roles in multiple plant developmental processes. Spikelets are the primary sink tissues during reproductive growth, and N deficiency can cause floral abortion. However, the roles of N nutrition in meiosis, the crucial step in plant sexual reproduction, are poorly understood. Here, we identified an N-dependent meiotic entrance mutant with loss of function of ELECTRON TRANSFER FLAVOPROTEIN SUBUNIT β (ETFβ) in rice (Oryza sativa). etfβ displayed meiosis initiation defects, excessive accumulation of branched-chain amino acids (BCAAs) and decrease in total N contents in spikelets under N starvation, which were rescued by applying excess exogenous inorganic N. Under N starvation, ETFβ, through its involvement in BCAA catabolism, promotes N reutilization and contributes to meeting N demands of spikelets, highlighting the impact of N nutrition on meiosis initiation. We conclude that N nutrition contributes to plant fertility by affecting meiosis initiation.
Product Description
Roche
DIG Nucleic Acid Detection Kit, sufficient for 40 blots (10 cm x 10 cm each), kit of 1 (5 components), suitable for hybridization, suitable for Northern blotting