- Ulvan-chitosan polyelectrolyte complexes as matrices for enzyme induced biomimetic mineralization.
Ulvan-chitosan polyelectrolyte complexes as matrices for enzyme induced biomimetic mineralization.
Carbohydrate polymers (2017-12-28)
Mamoni Dash, Sangram K Samal, Andrea Morelli, Cristina Bartoli, Heidi A Declercq, Timothy E L Douglas, Peter Dubruel, Federica Chiellini
PMID29279122
ABSTRACT
Polyelectrolyte complexes (PEC) of chitosan and ulvan were fabricated to study alkaline phosphatase (ALP) mediated formation of apatitic minerals. Scaffolds of the PEC were subjected to ALP and successful mineral formation was studied using SEM, Raman and XRD techniques. Investigation of the morphology via SEM shows globular structures of the deposited minerals, which promoted cell attachment, proliferation and extracellular matrix formation. The PEC and their successful calcium phosphate based mineralization offers a greener route of scaffold fabrication towards developing resorbable materials for tissue engineering.
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, lyophilized powder, ≥10 DEA units/mg solid