MilliporeSigma
  • Propeptide glycosylation and galectin-3 binding decrease proteolytic activation of human proMMP-9/progelatinase B.
Propeptide glycosylation and galectin-3 binding decrease proteolytic activation of human proMMP-9/progelatinase B.
The FEBS journal (2018-11-14)
Lise Boon, Estefania Ugarte-Berzal, Erik Martens, Jennifer Vandooren, Vasily Rybakin, Didier Colau, Monica Gordon-Alonso, Pierre van der Bruggen, Walter Stöcker, Christoph Becker-Pauly, Peter Witters, Eva Morava, Jaak Jaeken, Paul Proost, Ghislain Opdenakker
ABSTRACT
Matrix metalloproteinases (MMPs) are secreted as proenzymes, containing propeptides that interact with the catalytic zinc, thereby controlling MMP activation. The MMP-9 propeptide is unique in the MMP family because of its post-translational modification with an N-linked oligosaccharide. ProMMP-9 activation by MMP-3 occurs stepwise by cleavage of the propeptide in an aminoterminal (pro-AT) and carboxyterminal (pro-CT) peptide. We chemically synthesized aglycosyl pro-AT and pro-CT and purified recombinant glycosylated pro-ATS f-9 . First, we report new cleavage sites in the MMP-9 propeptide by MMP-3 and neutrophil elastase. Additionally, we demonstrated with the use of western blot analysis a higher resistance of glycosylated versus aglycosyl pro-AT against proteolysis by MMP-3, MMP-9, meprin α, neutrophil elastase and by protease-rich synovial fluids from rheumatoid arthritis patients. Moreover, we investigated the effect of glycosylation on proteolytic activation of human proMMP-9 with the use of zymography and dye-quenched gelatin cleavage analysis. Compared to recombinant Sf-9 proMMP-9 glycoforms, larger oligosaccharides of human neutrophil proMMP-9 increased resistance against proteolytic activation. Additionally, proMMP-9 from Congenital Disorder of Glycosylation patients, compared to healthy controls, showed a higher activation rate by MMP-3. Finally, we demonstrated that glycan-galectin-3 interactions reduced proMMP-9 activation. In conclusion, modification of MMP-9 propeptide glycosylation is a fine-tuning mechanism and co-determines the specific activity of MMP-9 in physiology and pathology. ENZYMES: MMP-9 EC 3.4.24.35, MMP-3 EC 3.4.24.17, meprin α EC 3.4.24.18, neutrophil elastase EC 3.4.21.37, trypsin EC 3.4.21.4 and PNGase F EC 3.5.1.52.
Sigma-Aldrich
MMP-3, Catalytic Domain, Human, Recombinant, E. coli